Learn More
ACROBiosystems Recombinant Protein;Human IL-15 Protein, Tag Free;E.coli;20UG;IL5,IL-15,IL15,Interleukin 15,Interleukin-15,Interleukin15,MGC9721
Supplier:  ACROBiosystems IL5H411720UG
Encompass_Preferred
Recombinant Protein;Human IL-15 Protein, Tag Free;20UG;Human IL-15, Tag Free (IL5-H4117) is expressed from E.coli cells. It contains AA Asn 49 - Ser 162 (Accession # AAH18149).;This protein carries no "tag". The protein has a calculated MW of 12.8 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IL-15, Tag Free (Cat. No. IL5-H4117) at 10μg/mL (100 μL/well) can bind Human IL-2 R beta, Fc Tag (Cat. No. ILB-H5253) with a linear range of 0.08-0.3 μg/mL (QC tested).;Human IL-15, Tag Free (Cat. No. IL5-H4117) stimulates proliferation of CTLL-2 cells. The ED50 for this effect is 0.097-0.190 ng/ml (Routinely tested).
Catalog No. 50-165-7442
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.