Learn More
ACROBiosystems Recombinant Protein;Human Leptin R / CD295 Protein, Fc Tag;HEK293;100UG;LER,LEPR,CD295,DKFZp686B1731,LEP-R,OB-R,OBR,Leptin receptor
Supplier:  ACROBiosystems LERH5252100UG
Encompass_Preferred
Recombinant Protein;Human Leptin R / CD295 Protein, Fc Tag;100UG;Human Leptin R, Fc Tag (LER-H5252) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Asp 839 (Accession # NP_002294.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 121 kDa. The protein migrates as 150-165 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7695
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.