Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD155 / PVR Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;CD5,CD155,PVR,FLJ25946,PVS,TAGE4,HVED,NECL5,NECL-5,TAGE-4
Supplier:  ACROBiosystems CD5H82E325UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD155 / PVR Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CD155, His Tag, Avi Tag (CD5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession # NP_006496.4).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 38.8 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIGIT, mouse IgG2a Fc tag, low endotoxin (Cat. No. TIT-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD155, His Tag, Avi Tag (Cat. No. CD5-H82E3) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Biotinylated Human CD155, His Tag, Avi Tag (Cat. No. CD5-H82E3) at 5 μg/mL (100 μL/well) via streptavidin can bind Human TIGIT Protein, Fc Tag (Cat. No. TIT-H5254) with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-165-8015
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.