Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc Tag, Avi Tag (Avitag™);HEK293;25UG;AN7,ANGPTL7,CDT6
Supplier: ACROBiosystems AN7H82F325UG
Recombinant Protein;Biotinylated Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human ANGPTL7, Mouse IgG2a Fc Tag, Avi Tag (AN7-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pro 346 (Accession # O43827-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 66.5 kDa. The protein migrates as 66 kDa and 75-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LILRB4, Fc Tag (Cat. No. LI4-H5259) at 1 μg/mL (100 μL/well) can bind Biotinylated Human ANGPTL7, Mouse IgG2a Fc Tag, Avi Tag (Cat. No. AN7-H82F3) with a linear range of 0.078-2.5 μg/mL (QC tested).;Immobilized Human LILRB2, Fc Tag (Cat. No. CDD-H5259) at 1 μg/mL (100 μL/well) can bind Biotinylated Human ANGPTL7, Mouse IgG2a Fc Tag, Avi Tag (Cat. No. AN7-H82F3) with a linear range of 0.078-1.25 μg/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.