Learn More
ACROBiosystems Recombinant Protein;Human PROCR / EPCR / CD201 Protein, Fc Tag;HEK293;100UG;PRR,PROCR,EPCR,CD201,APC receptor,CCCA,CCD41
Supplier: ACROBiosystems PRRH5254100UG
Recombinant Protein;Human PROCR / EPCR / CD201 Protein, Fc Tag;100UG;Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Thr 209 (Accession # AAH14451).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. As a result of glycosylation, the protein migrates as 65 kDa under reducing (R) condition, and 125 kDa under non-reducing (NR) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.