Learn More
ACROBiosystems Recombinant Protein;Human Integrin isoform alpha-7X2B beta 1 (ITGA7X2B&ITGB1) Heterodimer Protein;HEK293;100UG;IT1,Integrin isoform alpha-7X2B beta 1,ITGA7X2BB1,ITGA7X2B&ITGB1,ITGA7
Supplier:  ACROBiosystems IT1H52W8100UG
Encompass_Preferred
Recombinant Protein;Human Integrin isoform alpha-7X2B beta 1 (ITGA7X2B&ITGB1) Heterodimer Protein;100UG;Human ITGA7X2BB1, His Tag (IT1-H52W8) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Trp 1038 (ITGA7X2B) & Gln 21 - Asp 728 (ITGB1) (Accession # Q13683-7 (ITGA7X2B) & P05556-1 (ITGB1)).;Human ITGA7X2B & ITGB1 Heterodimer Protein, produced by co-expression of ITGA7X2B and ITGB1, has a calculated MW of 116.5 kDa (ITGA7X2B) and 83.7 kDa (ITGB1). Subunit ITGA7X2B is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 34 (ITGA7X2B) & Gln 21 (ITGB1). As a result of glycosylation, the protein migrates as 110 kDa (ITGA7X2B) and 120-140 kDa (ITGB1) under reducing (R) condition, and 115-125 kDa and 125-135 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8347
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.