Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human OX40 / TNFRSF4 / CD134 Protein (recommended for biopanning), Avi Tag (Avitag™);HEK293;25UG;TN4,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Supplier:  ACROBiosystems TN4H82E425UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human OX40 / TNFRSF4 / CD134 Protein (recommended for biopanning), Avi Tag (Avitag™);25UG;Biotinylated Human OX40, His Tag (TN4-H82E4) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # NP_003318.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 22.8 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-OX40 MAb, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human OX40, His Tag, Avi Tag (Cat. No. TN4-H82E4) with a linear range of 1-5 ng/mL (QC tested).;FACS assay shows that recombinant Biotinylated Human OX40, His Tag, Avi Tag (Cat. No. TN4-H82E4) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 is 0.1 μg/ml (Routinely tested).
Catalog No. 50-165-8437
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.