Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse IgG2a Fc, Avi Tag (Avitag™);HEK293;25UG;IGA,IgG2A Fc,IgG1 Fc,IgG2,IgG3,IgG4,IgG2a,IgG Fc,IgG1,IgG2a Fc
Supplier:  ACROBiosystems IGAM821025UG
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse IgG2a Fc, Avi Tag (Avitag™);25UG;Biotinylated Mouse IgG Fc, Avi Tag (IGA-M8210) is expressed from human 293 cells (HEK293). It contains AA Glu 98 - Lys 330 (Accession # P01863).;This protein carries an Avi tag (Avitag™) at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 36 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Recombinant Protein A at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse IgG2a Fc (Cat. No. IGA-M8210) with a linear range of 0.4-12.5 ng/mL (QC tested).
Catalog No. 50-165-9142
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.