Learn More
Sino Biological Mouse CD1D / R3G1 Protein (His & Fc Tag) 100ug
Supplier:  Sino Biological 50188M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD1D (NP_031665.2) (Met 1-Gly 305) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1539
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.