Learn More
Medchemexpress LLC HY-101582 5mg Medchemexpress, ZT 52656A hydrochloride CAS:115730-24-0 Purity:>98%
Supplier: Medchemexpress LLC HY1015825MG
Medchemexpress, HY-101582 5mg ZT 52656A hydrochloride CAS:115730-24-0 ZT 52656A is a selective kappa opioid agonist, used for the prevention or alleviation of pain in the eye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.