Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD27 Ligand / CD70 Protein, His,Avitag™,Flag Tag (active trimer) (MALS verified)
Supplier:  ACROBiosystems CDLH82D725UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD27 Ligand, His,Avitag,Flag Tag (active trimer) (MALS verified) (CDL-H82D7) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™) and a flag tag. The protein has a calculated MW of 51.0 kDa. The protein migrates as 47-57 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27, Fc Tag (Cat. No. CD7-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, His,Avitag,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H82D7) with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-195-6423
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.