Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque FGL1 Protein, Fc Tag
Supplier:  ACROBiosystems FG1C5269100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque FGL1, Fc Tag (FG1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # G7N0K6-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Mouse IgG2a Fc Tag (Cat. No. LA3-H52Aa) at 10 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque FGL1, Fc Tag (Cat. No. FG1-C5269) with a linear range of 0.01-0.313 μg/mL (QC tested).
Catalog No. 50-195-6532
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.