Learn More
ACROBiosystems ACROBiosystems Mouse B7-H2 / ICOSLG Protein, Fc Tag
Supplier: ACROBiosystems B72M5258100UG
Recombinant Protein;100UG;Mouse B7-H2, Fc Tag (B72-M5258) is expressed from human 293 cells (HEK293). It contains AA Glu 47 - Lys 279 (Accession # Q9JHJ8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.7 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse ICOS (C137S, C138S), His Tag (Cat. No. ICS-M52H8) at 5 μg/mL (100 μL/well) can bind Mouse B7-H2, Fc Tag (Cat. No. B72-M5258) with a linear range of 2-31 ng/mL (QC tested).