Learn More
ACROBiosystems ACROBiosystems Cynomolgus Complement C5 Protein, His Tag
Supplier:  ACROBiosystems CO5C52HX100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Complement C5, His Tag (CO5-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accession # XP_005580972.1).;This protein carries a polyhistidine tag at the C-terminus.The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 73.9 kDa (β chain) and 114.9 kDa (α chain). The protein migrates as 70 kDa (β chain) and 116 kDa (α chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Complement C5, His Tag (Cat. No. CO5-C52Hx) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-195-6832
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.