Learn More
ACROBiosystems ACROBiosystems Human IL-33 Protein, His Tag
Supplier: ACROBiosystems IL3H52H750UG
Recombinant Protein;50UG;Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession # O95760-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) at 5 μg/mL (100 μL/well) can bind Human IL-33, His Tag (Cat. No. IL3-H52H7) with a linear range of 5-20 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.