Learn More
ACROBiosystems ACROBiosystems Human IgG2 Fc Protein, Tag Free
Supplier: ACROBiosystems IG2H5206200UG
Recombinant Protein;200UG;Human IgG Fc, Tag Free (IG2-H5206) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 326 (Accession # P01859-1).;This protein carries no "tag". The protein has a calculated MW of 25.7 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.