Learn More
ACROBiosystems ACROBiosystems Anti-Cetuximab Antibodies (Non-Neutralizing)
Supplier:  ACROBiosystems CEBY31100UG
Encompass_Preferred
Antibody;100UG;IgG1/kappa;Recognizes Cetuximab specifically, no cross reactivity with other humanized antibodies.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab')2.; ;Anti-Cetuximab Antibodies bridging ELISA for Anti-Drug Antibody (ADA) assay development.Immobilized cetuximab at 1 µg/mL, add increasing concentrations of Anti-Cetuximab Antibodies (Cat. No. CEB-Y31, 10% human serum) and then add biotinylated cetuximab at 2 µg/mL. Detection was performed using HRP-conjugated streptavidin with a sensitivity of 39 ng/mL. ;Demonstration of the specificity of Anti-Cetuximab Antibodies (Cat. No. CEB-Y31) to the cetuximab.;Anti-Cetuximab Antibodies (mouse IgG1, Cat. No. CEB-Y31) captured on CM5 chip via anti-mouse antibodies surface, can bind human cetuximab with an affinity constant of 0.421 nM.
Catalog No. 50-195-6993
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.