Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CEACAM-6 / CD66c Protein, His,Avitag™
Supplier:  ACROBiosystems CE6H82E725UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CEACAM-6, His,Avitag (CE6-H82E7) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Gly 320 (Accession # P40199-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.9 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CEACAM-8, His Tag (Cat. No. CE8-H5224) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-6, His,Avitag (Cat. No. CE6-H82E7) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-195-7092
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.