Learn More
ACROBiosystems ACROBiosystems Vaccinia Virus B18R Protein, His Tag
Supplier:  ACROBiosystems B1RH52H650UG
Encompass_Preferred
Recombinant Protein;50UG;Vaccinia Virus B18R, His Tag, His 20 - Glu 351 (Accession # P25213), was produced in human 293 cells (HEK293) at ACROBiosystems.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IFN-α1a at 2 μg/mL (100 μL/well) can bind Vaccinia Virus B18R, His Tag (Cat. No. B1R-H52H6) with a linear range of 0.8-6 ng/mL (QC tested).
Catalog No. 50-195-7266
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.