Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein, His Tag&Tag Free
Supplier: ACROBiosystems IT5H52W5100UG
Recombinant Protein;100UG;Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free (IT5-H52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 24 - Asn 719 (ITGB5) (Accession # NP_002201.1 (ITGAV) & NP_002204.2 (ITGB5)).;Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB5, has a calculated MW of 112.9 kDa (ITGAV) and 81.9 kDa (ITGB5). Subunit ITGAV is fused with polyhistidine tag at the C-terminus and followed by a acidic tail and subunit ITGB5 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 130-150 kDa (ITGAV) and 85-95 kDa (ITGB5) respectively due to glycosylation.;Immobilized Vitronectin at 2 μg/mL (100 μL/well) can bind Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT5-H52W5) with a linear range of 2-20 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.