Learn More
ACROBiosystems ACROBiosystems Human CX3CL1 / Fractalkine Protein, His Tag
Supplier:  ACROBiosystems CX1H522150UG
Encompass_Preferred
Recombinant Protein;50UG;Human CX3CL1, His Tag (CX1-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 341 (Accession # AAH01163).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7481