Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque TIGIT Protein, Fc Tag
Supplier: ACROBiosystems TITC525450UG
Recombinant Protein;50UG;Cynomolgus / Rhesus macaque TIGIT, Fc Tag (TIT-C5254) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # G7NXM4-1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.9 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD155, His Tag at 10 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque TIGIT, Fc Tag (Cat. No. TIT-C5254) with a linear range of 2.22-20 μg/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.