Learn More
ACROBiosystems Human BAFFR / TNFRSF13C Protein, His Tag
Supplier: ACROBiosystems BARH52H3100UG
Recombinant Protein;100UG;Human BAFFR Protein, His Tag (BAR-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 8.4 kDa.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.