Learn More
ACROBiosystems Biotinylated Human CD229 / SLAMF3 Protein, His,Avitag™
Supplier:  ACROBiosystems CD9H82E625UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD229, His,Avitag™ (CD9-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 48 - Lys 454 (Accession # Q9HBG7-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 48.4 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-201-9495
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.