Learn More
ACROBiosystems Cynomolgus Latent TGF-beta 1 (C33S) Protein, His Tag
Supplier:  ACROBiosystems TG1C524350UG
Encompass_Preferred
Recombinant Protein;50UG;Cynomolgus Latent TGF-Beta 1 (C33S), His Tag (TG1-C5243) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (C33S) (Accession # A0A2K5TJB2-1 (C33S)).;This protein carries a polyhistidine tag at the N-terminus. The precursor proprotein is partially cleaved by FURIN to form mature Transforming growth factor beta-1 (TGF-beta-1) and Latency-associated peptide (LAP) chains with calculated MW of 12.8 kDa and 30.4 kDa respectively. The protein migrates as 50 kDa (precursor proprotein), 37-45 kDa (LAP) and 13 kDa (TGFB1) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Latent TGF-Beta 1 (C33S), His Tag (Cat. No. TG1-C5243) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 0.6-20 ng/mL (QC tested).;Immobilized Cynomolgus Latent TGF-Beta 1 (C33S), His Tag (Cat. No. TG1-C5243) at 2 μg/mL (100 μL/well) can bind Bio
Catalog No. 50-201-9551
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.