Learn More
ACROBiosystems Human IFN-alpha 1 Protein, Fc Tag
Supplier: ACROBiosystems IFAH5258100UG
Recombinant Protein;100UG;Human IFN-alpha 1, Fc Tag (IFA-H5258) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 189 (Accession # P01562-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.8 kDa.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.