Learn More
ACROBiosystems Biotinylated Human CD19 (20-291) Protein, His,Avitag™ (SPR verified)
Supplier: ACROBiosystems CD9H82E925UG
Recombinant Protein;25UG;Biotinylated Human CD19 (20-291), His,Avitag (CD9-H82E9) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession # P15391-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 33.7 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human CD19 Antibody, Mouse IgG2a at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD19 (20-291), His,Avitag (Cat. No. CD9-H82E9) with a linear range of 1-31 ng/mL (Routinely tested).;Immobilized Biotinylated Human CD19 (20-291), His,Avitag (Cat. No. CD9-H82E9) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human CD19 Antibody, Mouse IgG2a with a linear range of 0.1-6 ng/mL (Routinely tested).;Biotinylated Human CD19 (20-291), His,Avitag (Cat. No. CD9-H82E9) captured on Biotin CAP - Ser
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.