Learn More
ACROBiosystems ActiveMax Human CCL19 / MIP-3 beta Protein, Tag Free
Supplier:  ACROBiosystems CC9H521550UG
Encompass_Preferred
Recombinant Protein;50UG;ActiveMax Human CCL19 / MIP-3 beta, Tag Free (CC9-H5215) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Ser 98 (Accession # Q99731-1).;This protein carries no "tag". The protein has a calculated MW of 8.8 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9775
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.