Learn More
ACROBiosystems Human CD3 gamma / CD3G Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems CDAH5254100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD3 gamma, Fc Tag (CDA-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 116 (Accession # P09693-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4759
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.