Learn More
ACROBiosystems Biotinylated Human PSCA Protein, His,Avitag™
Supplier:  ACROBiosystems PSAH82E325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human PSCA, His,Avitag (PSA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 12 - Ser 86 (Accession # O43653-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.1 kDa.
Catalog No. 50-205-4761
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.