Learn More
ACROBiosystems Biotinylated Human PTK7 / CCK4 protein, His,Avitag™
Supplier: ACROBiosystems PT7H82E5200UG
Recombinant Protein;200UG;Biotinylated Human PTK7, His,Avitag (PT7-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 31 - Thr 704 (Accession # Q13308-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 78.2 kDa.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.