Learn More
ACROBiosystems Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (AS86)
Supplier:  ACROBiosystems S2NS86100UG
Encompass_Preferred
Antibody;100UG;Human IgG4/kappa;This product is a specific antibody against SARS-CoV-2 Spike S2 protein. Cross-reactivity with S2 protein of other coronaviruses has not been tested.;Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (AS86) is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from CHO cells.; The protein migrates as 27 kDa and 55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 S2 protein, His Tag (Cat. No. S2N-C52H5) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (Cat. No. S2N-S86) with a linear range of 0.12-2 ng/mL (QC tested).
Catalog No. 50-205-4830
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.