Learn More
ACROBiosystems Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag™ (Y45)
Supplier:  ACROBiosystems FM3BY5425TESTS
Encompass_Preferred
Antibody;25TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag (Cat. No. FM3-BY54) is expressed from human 293 cells.; ;2e5 of Anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag (Cat. No. FM3-BY54) and isotype control respectively, washed and then followed by PE-SA and analyzed with FACS (QC tested).
Catalog No. 50-205-4870
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.