Learn More
ACROBiosystems FITC-Labeled Human CD72 Protein, Fc Tag
Supplier:  ACROBiosystems CD2HF25825UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human CD72, Fc Tag (CD2-HF258) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). It is the FITC labeled form of Human CD72, Fc Tag (Cat. No.CD2-H5251).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.2 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5123
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.