Learn More
ACROBiosystems HCoV-NL63 Nucleocapsid protein, His Tag
Supplier:  ACROBiosystems NUNV5146100UG
Encompass_Preferred
Recombinant Protein;100UG;HCoV-NL63 Nucleocapsid, His Tag (NUN-V5146) is expressed from E.coli cells. It contains AA Met 1 - His 377 (Accession # Q6Q1R8-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 44 kDa.
Catalog No. 50-205-5133
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.