Learn More
Description
Includes
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
Specifications
| MWCO | 8 to 10 kDa |
| Diameter (Metric) | 6.4 mm |
| Length (Metric) | 0.32 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
