Learn More
Advanced Biomatrix Irgacure 2959 Photoinitiator 5 grams non-sterile
Supplier:  Advanced Biomatrix 52005GM
Encompass_Preferred
Irgacure 2959 is a highly efficient non-yellowing radical photoinitiator for the UV curing of systems, such as hydrogels and bioinks. Irgacure is most efficiently activated using 365nm wavelength UV light. Irgacure must be solubilized in methanol, as it is not water soluble (such as LAP and Ruthenium).
Catalog No. 50-225-5918