Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human HLA-A*11:01 & B2M complex with KRAS G12V peptide, His-AviTag, 25 μg

Supplier:  ACROBiosystems HLVH82E625UG

Encompass_Preferred

ACROBiosystems biotinylated complex of human HLA-A*11:01, beta-2 microglobulin (B2M), and KRAS G12V peptide (VVGAVGVGK). Expressed in HEK293 cells and affinity-tagged for purification with site-specific biotin labeling. Supplied lyophilized in a 25 μg pack for use in binding assays, assay development, and biophysical characterization.

  • Species: Human.
  • Expression system: HEK293 cells.
  • Composition: HLA-A*11:01, B2M, KRAS G12V peptide.
  • Tags and label: His tag, AviTag, biotin-labeled.
  • Format and pack size: Lyophilized powder, 25 μg.
  • Storage: Store at -20 °C; ship at room temperature.
  • Applications: Binding studies, assay development, biophysical analysis.

Catalog No. 50-237-0217


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.