Learn More
ACROBiosystems Cynomolgus Carbonic Anhydrase IX (CA9) protein, His-tag, 50 μg
Supplier: ACROBiosystems CA9C52H350UG
Recombinant cynomolgus carbonic anhydrase IX (CA9) is supplied as a lyophilized, C-terminal His-tagged purified protein fragment spanning Gln 38-Leu 398. The protein is glycosylated and characterized by SEC-MALS, SDS-PAGE, and SPR to confirm purity and activity, making it suitable for biochemical characterization and binding studies.
- Species: Cynomolgus
- Tag: C-terminal polyhistidine (His) tag
- Unit size: 50 μg
- Residues: Gln 38-Leu 398
- Molecular weight: Calculated 41.5 kDa; migrates 49-53 kDa by SDS-PAGE
- Purity and activity: MALS verified (>90% purity), SDS-PAGE (>95% purity), SPR KD ~27.6 nM, specific activity >55 pmol/min/μg
- Formulation: Lyophilized from 25 mM MES, 100 mM NaCl, pH 6.5, with trehalose
- Applications: Biochemical assays, binding and affinity studies, SPR, SEC-MALS
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.