Learn More
Sino Biological Mouse TNFRSF1B PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40589V08H42100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TNFR2 / CD120b / TNFRSF1B (rM TNFR2 / CD120b / TNFRSF1B; Catalog#50128-M08H; NP_035740.2; Met 1-Gly 258). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4320
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.