Learn More
Teknova Brain Heart Infusion Agar Plates with Streptomycin-50. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
Supplier: Teknova B11254PK
3.7% Brain Heart Infusion Broth,1.5% Agar,50µg/mL Streptomycin Sulfate