Learn More
Teknova Pseudomonas Isolation Agar Plates (PIA). 100mm Plates, 20 Plates/Sleeve, Sterile.
Supplier: Teknova P0144
2.0% Tryptone, 0.14% Magnesium Chloride,1.0% Potassium Sulfate, 0.003% Irgasan, 2.0% Glycerol, 1.36% Agar
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.