Learn More
Teknova YPD Agar Plates. 40mL Fill, 100mm Plates, 20 Plates/Sleeve, Sterile.
Supplier: Teknova Y1030
1.0% Yeast Extract, 2.0% Peptone, 2.0% Glucose, 2.0% Agar
This item is not returnable.
View return policy