Learn More
Immunology Consultants Mouse IgE ELISA Kit
SDP
Supplier:  Immunology Consultants E90E
Encompass_Preferred
The Mouse IgE test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgE in mouse biological samples.The assay is a double antibody sandwich Mouse IgE ELISA.
Catalog No. 50-240-6592
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.