Learn More
ACROBiosystems SARS-CoV-2 Spike RBD 25ug
Supplier:  ACROBiosystems SPDC82Q425UG
Encompass_Preferred
Biotinylated SARS-CoV-2 Spike RBD Protein HisAvitag (XBB.1.16/Omicron) (SPD-C82Q4) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339H R346T L368I S371F S373P S375F T376A D405N R408S K417N N440K V445P G446S N460K S477N T478R E484A F486P F490S Q498R N501Y Y505H)). The spike mutations are identified on the SARS-CoV-2 Omicron variant (Pango lineage: XBB.1.16).
Catalog No. 50-253-4152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.