Learn More
ACROBiosystems Mouse LIV-1 Protein Fc Tag
Supplier:  ACROBiosystems LV1M52531MG
Encompass_Preferred
Mouse LIV-1 Protein Fc Tag (LV1-M5253) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Trp 335 (Accession # Q8C145-1).
Catalog No. 50-253-4492
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.