Learn More
ACROBiosystems Human HLA-A0206 B2M 25ug
Supplier: ACROBiosystems HLMH82EP25UG
Biotinylated Human HLA-A*0206B2M Monomer Protein (HLM-H82Ep) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*0206) Ile 21 - Met 119 (B2M) (Accession # AAW30168.1 (HLA-A*0206) P61769 (B2M)).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.