Learn More
ACROBiosystems Biotinylated Human HLA-A0201
Supplier: ACROBiosystems HLEH82E825UG
Biotinylated Human HLA-A*0201B2MPRAME (SLLQHLIGL) Complex Protein (HLE-H82E8) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (B2M) Gly 25 - Ile 308 (HLA-A*0201) SLLQHLIGL peptide (Accession # P61769 (B2M) AAA59606.1 (HLA-A*0201) SLLQHLIGL).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.