Learn More
ACROBiosystems Mouse GUCY2C Protein His Tag
Supplier:  ACROBiosystems GUCM53H5100UG
Encompass_Preferred
Mouse GUCY2C Protein His Tag (GUC-M53H5) is expressed from CHO cells. It contains AA Val 20 - Met 433 (Accession # Q3UWA6-1).
Catalog No. 50-253-4921
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.