Learn More
ACROBiosystems Human GUCY2C 25tests
Supplier:  ACROBiosystems GUCHA2H425TESTS
Encompass_Preferred
APC-Labeled Human GUCY2C Protein His Tag (GUC-HA2H4) is produced via conjugation of APC to Human GUCY2C Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human GUCY2C Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).
Catalog No. 50-253-4934
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.